Anti-CLASP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: United States Biological
USBIC5831-01F-PEEA 702 EUR
USBIC5831-01F-PE
Anti-CLASP1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: CLASP1 (cytoplasmic linker associated protein 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR