Anti-NLRC4 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC5820-01CEA 419 EUR
USBIC5820-01C
Anti-NLRC4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 1:1000-1:5000; Immunoprecipitation: 1:50-1:200

Type: Primary
Antigen: NLRC4 (NLR family, CARD domain containing 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR