Anti-JUN Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC5815-10UEA 347 EUR
USBIC5815-10U
Anti-JUN Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR