Anti-TRAF3IP2 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2245]

USBIC5072-75H-PEEA 0 EUR
USBIC5072-75H-PE
Anti-TRAF3IP2 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 10B2245]
Type: Primary
Antigen: TRAF3IP2 (TRAF3 interacting protein 2)
Clonality: Monoclonal
Clone: 10B2245
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR