Anti-CIDEC Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC5072-57EA 473 EUR
USBIC5072-57
Anti-CIDEC Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1000

Type: Primary
Antigen: CIDEC (cell death-inducing DFFA-like effector c)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR