Anti-CIDEA Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC5072-03CEA 616 EUR
USBIC5072-03C
Anti-CIDEA Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CIDEA (cell death-inducing DFFA-like effector a)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR