Anti-SLC5A7 Mouse Monoclonal Antibody [clone: 5K101]

Lieferant: United States Biological
USBIC5071-50EA 473 EUR
USBIC5071-50
Anti-SLC5A7 Mouse Monoclonal Antibody [clone: 5K101]
Antikörper
Type: Primary
Antigen: SLC5A7 (solute carrier family 5 (sodium/choline cotransporter), member 7)
Clonality: Monoclonal
Clone: 5K101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR