Anti-C7orf50 Goat polyclonal antibody

Lieferant: United States Biological
USBIC5071-07CEA 420 EUR
USBIC5071-07C
Anti-C7orf50 Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1: 32000

Type: Primary
Antigen: C7orf50 (chromosome 7 open reading frame 50)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR