Anti-Pancreas Mouse Monoclonal Antibody [clone: 10B2241]

Lieferant: United States Biological
USBIC5070-01DEA 599 EUR
USBIC5070-01D
Anti-Pancreas Mouse Monoclonal Antibody [clone: 10B2241]
Antikörper
Type: Primary
Antigen: Pancreas
Clonality: Monoclonal
Clone: 10B2241
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR