Anti-CHGA Mouse Monoclonal Antibody

Lieferant: United States Biological
USBIC5069-96ZEA 633 EUR
USBIC5069-96Z
Anti-CHGA Mouse Monoclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:2000; Immunochemistry: 1:200

Type: Primary
Antigen: CHGA (chromogranin A (parathyroid secretory protein 1))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR