Anti-Chorionic Gonadotropin, Human Chicken Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC5069-75CEA 407 EUR
USBIC5069-75C
Anti-Chorionic Gonadotropin, Human Chicken Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.1-0.5 ?g/ml; Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: Chorionic Gonadotropin, Human
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR