Anti-Chorionic Gonadotropin, Human Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC5069-17DEA 411 EUR
USBIC5069-17D
Anti-Chorionic Gonadotropin, Human Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Chorionic Gonadotropin, Human
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR