Anti-CHRD Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC5068-60AEA 527 EUR
USBIC5068-60A
Anti-CHRD Rabbit polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: CHRD (chordin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR