Anti-Chondroitin Sulfate Proteoglycan Mouse Monoclonal Antibody [clone: 5H26]

Lieferant: United States Biological
USBIC5067-71EA 686 EUR
USBIC5067-71
Anti-Chondroitin Sulfate Proteoglycan Mouse Monoclonal Antibody [clone: 5H26]
Antikörper
Recommended dilution: Western Blot: 0.25-0.5 ?g/ml

Type: Primary
Antigen: Chondroitin Sulfate Proteoglycan
Clonality: Monoclonal
Clone: 5H26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR