Anti-DDIT3 Mouse monoclonal antibody [clone: 6D313]

Lieferant: United States Biological
USBIC5064-05EA 776 EUR
USBIC5064-05
Anti-DDIT3 Mouse monoclonal antibody [clone: 6D313]
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: DDIT3 (DNA-damage-inducible transcript 3)
Clonality: Monoclonal
Clone: 6D313
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR