Anti-CHKA Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC5062-69A-BIOTEA 852 EUR
USBIC5062-69A-BIOT
Anti-CHKA Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CHKA (choline kinase alpha)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR