Anti-CCKBR Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC5042-25EEA 703 EUR
USBIC5042-25E
Anti-CCKBR Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: CCKBR (cholecystokinin B receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR