Anti-CCKAR Goat polyclonal antibody

Lieferant: United States Biological
USBIC5042-20HEA 471 EUR
USBIC5042-20H
Anti-CCKAR Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:64000

Type: Primary
Antigen: CCKAR (cholecystokinin A receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR