Anti-CCK Rabbit Polyclonal Antibody (Cy5®)

Lieferant: United States Biological
USBIC5042-01M4EA 923 EUR
USBIC5042-01M4
Anti-CCK Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Type: Primary
Antigen: CCK (cholecystokinin)
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR