Anti-Cholera Toxin, beta Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: United States Biological
USBIC4275-35QEA 416 EUR
USBIC4275-35Q
Anti-Cholera Toxin, beta Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: Immunochemistry: 1: 20-1: 200; Western Blot: 1: 20-1: 200; ELISA: 1: 200-1: 000

Type: Primary
Antigen: Cholera Toxin, beta
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR