Anti-Chlamydia trachomatis L2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC4250-52GEA 383 EUR
USBIC4250-52G
Anti-Chlamydia trachomatis L2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Chlamydia trachomatis L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR