Anti-Chlamydia trachomatis L2 Goat Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC4250-52BEA 473 EUR
USBIC4250-52B
Anti-Chlamydia trachomatis L2 Goat Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Chlamydia trachomatis L2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR