Anti-Chlamydia trachomatis Mouse Monoclonal Antibody [clone: 3]

Lieferant: United States Biological
USBIC4250-04GEA 608 EUR
USBIC4250-04G
Anti-Chlamydia trachomatis Mouse Monoclonal Antibody [clone: 3]
Antikörper
Recommended dilution: Immunochemistry: 1:20

Type: Primary
Antigen: Chlamydia trachomatis
Clonality: Monoclonal
Clone: 3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR