Anti-CHEK2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC4200-90PEA 578 EUR
USBIC4200-90P
Anti-CHEK2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR