Anti-CHEK1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: United States Biological
USBIC4200-05H-APEA 720 EUR
USBIC4200-05H-AP
Anti-CHEK1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Dot Blot: 1:500
Do not freeze!

Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR