Anti-CHEK1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC4200-05EEA 357 EUR
USBIC4200-05E
Anti-CHEK1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR