Anti-Chemokine Receptor FKSG80 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC3915-02G-BIOTEA 619 EUR
USBIC3915-02G-BIOT
Anti-Chemokine Receptor FKSG80 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Chemokine Receptor FKSG80
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR