Anti-Chemokine Receptor FKSG80 Rabbit polyclonal antibody HRP (Horseradish Peroxidase)

USBIC3915-02E-HRPEA 0 EUR
USBIC3915-02E-HRP
Anti-Chemokine Receptor FKSG80 Rabbit polyclonal antibody HRP (Horseradish Peroxidase)
Recommended dilution: ELISA: 1:1000; Western Blot: 1:60; Immunochemistry: 1:60

Type: Primary
Antigen: Chemokine Receptor FKSG80
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR