Anti-Chemerin Rat Monoclonal Antibody [clone: 9L816]

Lieferant: United States Biological
USBIC3890-01BEA 738 EUR
USBIC3890-01B
Anti-Chemerin Rat Monoclonal Antibody [clone: 9L816]
Antikörper
Recommended dilution: ELISA: 2-8?g/ml

Type: Primary
Antigen: Chemerin
Clonality: Monoclonal
Clone: 9L816
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR