Anti-CCT8L2 Mouse monoclonal antibody Biotin [clone: 10B2210]

USBIC3011-15-BIOTEA 0 EUR
USBIC3011-15-BIOT
Anti-CCT8L2 Mouse monoclonal antibody Biotin [clone: 10B2210]
Type: Primary
Antigen: CCT8L2 (chaperonin containing TCP1, subunit 8 (theta)-like 2)
Clonality: Monoclonal
Clone: 10B2210
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR