Anti-CP Sheep Polyclonal Antibody

Lieferant: United States Biological
USBIC2900-01JEA 0 EUR
USBIC2900-01J
Anti-CP Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CP (ceruloplasmin (ferroxidase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR