Anti-CP Chicken Polyclonal Antibody

Lieferant: United States Biological
USBIC2900-01EEA 285 EUR
USBIC2900-01E
Anti-CP Chicken Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:200-1:500; Western Blot: 1:1000

Type: Primary
Antigen: CP (ceruloplasmin (ferroxidase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR