Anti-CDNF Rat monoclonal antibody [clone: 12K60]

Lieferant: United States Biological
USBIC2795-50BEA 796 EUR
USBIC2795-50B
Anti-CDNF Rat monoclonal antibody [clone: 12K60]
Antikörper
Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: CDNF (cerebral dopamine neurotrophic factor)
Clonality: Monoclonal
Clone: 12K60
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR