Anti-CERK Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC2778-22EA 578 EUR
USBIC2778-22
Anti-CERK Rabbit polyclonal antibody
Antikörper
Recommended dilution: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: CERK (ceramide kinase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR