Anti-ADAP2 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC2617-02B3EA 471 EUR
USBIC2617-02B3
Anti-ADAP2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: ADAP2 (ArfGAP with dual PH domains 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR