Anti-CENPK Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10C7]

USBIC2615-24K-APCEA 0 EUR
USBIC2615-24K-APC
Anti-CENPK Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10C7]
Type: Primary
Antigen: CENPK (centromere protein K)
Clonality: Monoclonal
Clone: 10C7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR