Anti-Cellulase Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC2610-01AEA 683 EUR
USBIC2610-01A
Anti-Cellulase Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: Cellulase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR