Anti-CECR5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: United States Biological
USBIC2592-05-PEEA 702 EUR
USBIC2592-05-PE
Anti-CECR5 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: CECR5 (cat eye syndrome chromosome region, candidate 5)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR