Anti-CEBPA Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC2589-90GEA 411 EUR
USBIC2589-90G
Anti-CEBPA Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1-4?g/ml

Type: Primary
Antigen: CEBPA (CCAAT/enhancer binding protein (C/EBP), alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR