Anti-CEBPA Sheep polyclonal antibody

Lieferant: United States Biological
USBIC2589-90B1EA 640 EUR
USBIC2589-90B1
Anti-CEBPA Sheep polyclonal antibody
Antikörper
Recommended dilution: Immunochemistry: 5-15 ?g/ml

Type: Primary
Antigen: CEBPA (CCAAT/enhancer binding protein (C/EBP), alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR