Anti-CEACAM 20 Mouse monoclonal antibody [clone: 9E85]

Lieferant: United States Biological
USBIC2589-87GEA 681 EUR
USBIC2589-87G
Anti-CEACAM 20 Mouse monoclonal antibody [clone: 9E85]
Antikörper
Type: Primary
Antigen: CEACAM 20
Clonality: Monoclonal
Clone: 9E+85
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR