Anti-CEACAM-4 Mouse monoclonal antibody [clone: 11D36]

Lieferant: United States Biological
USBIC2589-69EA 578 EUR
USBIC2589-69
Anti-CEACAM-4 Mouse monoclonal antibody [clone: 11D36]
Antikörper
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 2?g/ml

Type: Primary
Antigen: CEACAM-4
Clonality: Monoclonal
Clone: 11D36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR