Anti-CDX2 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC2589-57CEA 426 EUR
USBIC2589-57C
Anti-CDX2 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1-3?g/ml

Type: Primary
Antigen: CDX2 (caudal type homeobox 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Horse, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR