Anti-CDw76 Mouse monoclonal antibody [clone: BL-B8]

Lieferant: United States Biological
USBIC2586-50EA 421 EUR
USBIC2586-50
Anti-CDw76 Mouse monoclonal antibody [clone: BL-B8]
Antikörper
Type: Primary
Antigen: CDw76
Clonality: Monoclonal
Clone: BL-B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR