Anti-ITGA6 Mouse Monoclonal Antibody [clone: BQ16]

Lieferant: United States Biological
USBIC2585-25EEA 473 EUR
USBIC2585-25E
Anti-ITGA6 Mouse Monoclonal Antibody [clone: BQ16]
Antikörper
Type: Primary
Antigen: ITGA6 (integrin, alpha 6)
Clonality: Monoclonal
Clone: BQ16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR