Anti-CDw17 Mouse monoclonal antibody [clone: 5F20]

Lieferant: United States Biological
USBIC2584-01EA 339 EUR
USBIC2584-01
Anti-CDw17 Mouse monoclonal antibody [clone: 5F20]
Antikörper
Type: Primary
Antigen: CDw17
Clonality: Monoclonal
Clone: 5F20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR