Anti-CDK5 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC2575-01BEA 480 EUR
USBIC2575-01B
Anti-CDK5 Rabbit polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:500-1:2000

Type: Primary
Antigen: CDK5 (cyclin-dependent kinase 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR