Anti-CDK4 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2179]

USBIC2574-06C-FITCEA 0 EUR
USBIC2574-06C-FITC
Anti-CDK4 Mouse monoclonal antibody FITC (Fluorescein) [clone: 10B2179]
Type: Primary
Antigen: CDK4 (cyclin-dependent kinase 4)
Clonality: Monoclonal
Clone: 10B2179
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR