Anti-CDK4 Mouse monoclonal antibody [clone: 10K148]

Lieferant: United States Biological
USBIC2574-05GEA 440 EUR
USBIC2574-05G
Anti-CDK4 Mouse monoclonal antibody [clone: 10K148]
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CDK4 (cyclin-dependent kinase 4)
Clonality: Monoclonal
Clone: 10K148
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR