Anti-CDK1 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC2571-07FEA 675 EUR
USBIC2571-07F
Anti-CDK1 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR